Human Recombinant WHSC1 (from Insect cells), Unconjugated
: Biortus
: Frozen liquid on dry ice.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt Q05CW4
- Protein/Peptide Name:WHSC1
- Purity:85%
- Sequence:Q2-K568
- Formulation:50 mM Tris-HCl (pH7.5), 200 mM NaCl, 20% glycerol, 1 mM DTT
- Shipping Temperature:Frozen liquid with dry ice
Frequently Bought Together