Human Recombinant UHRF2 (from E. coli), Unconjugated
: Biortus
: Frozen liquid on dry ice.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt Q96PU4
- Protein Synonyms:RING finger protein 107
- Protein/Peptide Name:UHRF2
- Purity:0.85
- Sequence:W2-R802
- Formulation:50 mM Tris-HCl (pH7.5), 200 mM NaCl, 20% glycerol, 1 mM DTT
- Shipping Temperature:Frozen liquid with dry ice
Frequently Bought Together