Human Recombinant RNF181 (from Insect cells), Unconjugated
: Biortus
: Frozen liquid on dry ice.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt Q9NV06
- Protein Synonyms:DDB1- and CUL4-associated factor 13|DCAF13
- Protein/Peptide Name:RNF181
- Purity:90%
- Sequence:K2-K445
- Formulation:50 mM HEPES (pH 7.5), 250 mM NaCl
- Shipping Temperature:Frozen liquid with dry ice
Frequently Bought Together