Human Recombinant RNF141 (from E. coli), Unconjugated
: Biortus
: Frozen liquid on dry ice.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt Q8WVD5
- Protein/Peptide Name:RNF141
- Purity:90%
- Sequence:G2-P230
- Formulation:50 mM Tris-HCl (pH 7.5), 250 mM NaCl
- Shipping Temperature:Frozen liquid with dry ice
Frequently Bought Together