Human Recombinant LONRF2 (from E. coli), Unconjugated
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt Q1L5Z9
- Protein Synonyms:RING finger protein 192
- Protein/Peptide Name:LONRF2
- Purity:85%
- Sequence:S2-N754
- Formulation:50 mM Tris-HCl (pH7.5), 150 mM NaCl, 1 mM DTT, 20% glycerol
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together