Order Entry
United States
ContactUsLinkComponent
Human Recombinant KLHL42 (from Insect cells), Unconjugated
Human Recombinant KLHL42 (from Insect cells), Unconjugated
Supplier:  Biortus
Certificates
undefined
Human Recombinant KLHL42 (from Insect cells), Unconjugated
Supplier:  Biortus

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Insect cells
  • Species:
    Human
  • Gene ID:
    UniProt Q9P2K6
  • Protein Synonyms:
    Cullin-3-binding protein 9|Ctb9
  • Protein/Peptide Name:
    KLHL42
  • Purity:
    90%
  • Sequence:
    S2-T505
  • Formulation:
    50 mM Tris-HCl (pH 7.5), 200 mM NaCl, 20% glycerol, 1 mM DTT
  • Shipping Temperature:
    Frozen liquid with dry ice

Specifications

Related Information

Frequently Bought Together