Human Recombinant KBTBD2 (from Insect cells), Unconjugated
: Biortus
: Frozen liquid on dry ice.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt Q8IY47
- Protein Synonyms:BTB and kelch domain-containing protein 1
- Protein/Peptide Name:KBTBD2
- Purity:80%
- Sequence:S2-V623
- Formulation:50 mM Tris-HCl (pH 7.5), 200 mM NaCl, 20% glycerol, 1 mM DTT
- Shipping Temperature:Frozen liquid with dry ice
Frequently Bought Together