Human Recombinant HLTF (from E. coli), Unconjugated
Supplier: Biortus
Certificates
About this item
Delivery information: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt Q14527
- Protein Synonyms:RING finger protein 80
- Protein/Peptide Name:HLTF
- Purity:90%
- Sequence:V58-A174
- Formulation:50 mM Tris-HCl (pH 7.5), 1 mM DTT, 200 mM NaCl, 20% glycerol
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together