SARS-CoV-2 Recombinant rep Protein (from E. coli), Unconjugated
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:SARS-CoV-2
- Gene ID:UniProt P0DTC1
- Protein/Peptide Name:rep
- Purity:90%
- Sequence:A1-Q139
- Formulation:10 mM Tris 8.0,150 mM NaCl
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together