SARS-CoV-2 Recombinant NCP-N Protein (from E. coli), His
: Biortus
: Frozen liquid on dry ice.
- Conjugation:His
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:SARS-CoV-2
- Gene ID:UniProt P0DTC9
- Protein/Peptide Name:NCP-N
- Purity:90%
- Sequence:M1-A419
- Formulation:50 mM Tris-HCl (pH8.0), 600 mM NaCl, 5% glycerol
- Shipping Temperature:Frozen liquid with dry ice
Frequently Bought Together