Pyrococcus furiosus Recombinant radA Protein (from E. coli), Unconjugated
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Pyrococcus furiosus
- Gene ID:UniProt O74036
- Protein Synonyms:RecA-like NTPase
- Protein/Peptide Name:radA
- Purity:90%
- Sequence:A108-D349 end
- Formulation:50 mM Tris-HCl (pH7.5), 200 mM NaCl, 1 mM DTT, 5% glycerol
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together