Order Entry
United States
ContactUsLinkComponent
Human Recombinant NRAS Protein (from E. coli), GST
Human Recombinant NRAS Protein (from E. coli), GST
Supplier:  Biortus
Certificates
undefined
Human Recombinant NRAS Protein (from E. coli), GST
Supplier:  Biortus

Specifications

  • Conjugation:
    GST
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Gene ID:
    UniProt P01111
  • Protein Synonyms:
    Ras|GTPase NRas
  • Protein/Peptide Name:
    NRAS
  • Purity:
    95%
  • Sequence:
    T2-K169, Q61L
  • Formulation:
    20 mM HEPES (pH 7.5), 10% glycerol, 150 mM NaCl.
  • Shipping Temperature:
    Frozen liquid with dry ice

Specifications

Frequently Bought Together