Human Recombinant NRAS Protein (from E. coli), GST
Supplier: Biortus
Certificates
About this item
Delivery information: Frozen liquid on dry ice.
Specifications
- Conjugation:GST
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt P01111
- Protein Synonyms:Ras|GTPase NRas
- Protein/Peptide Name:NRAS
- Purity:95%
- Sequence:T2-K169, Q61K
- Formulation:20 mM HEPES (pH 7.5), 10% glycerol, 150 mM NaCl.
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together