Order Entry
United States
ContactUsLinkComponent
Human Recombinant KRAS Protein (from E. coli), Unconjugated
Human Recombinant KRAS Protein (from E. coli), Unconjugated
Supplier:  Biortus
Certificates
undefined
Human Recombinant KRAS Protein (from E. coli), Unconjugated
Supplier:  Biortus

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Gene ID:
    UniProt P01116
  • Protein Synonyms:
    GTPase KRas|Ras
  • Protein/Peptide Name:
    KRAS
  • Purity:
    95%
  • Sequence:
    M1-K169
  • Formulation:
    50 mM HEPES (pH 7.4), 5 mM MgCl₂, 150 mM NaCl, 1 mM DTT
  • Shipping Temperature:
    Frozen liquid with dry ice

Specifications

Frequently Bought Together