Human Recombinant KRAS Protein (from E. coli), biotinylated
Supplier: Biortus
Certificates
About this item
Delivery information: Frozen liquid on dry ice.
Specifications
- Conjugation:Biotin
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt P01116
- Protein Synonyms:GTPase KRas|Ras
- Protein/Peptide Name:KRAS
- Purity:95%
- Sequence:M1-M188,G12C
- Formulation:20 mM HEPES (pH 7.5), 1 mM MgCl₂, 150 mM NaCl, 0.5 mM TCEP
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together