Order Entry
United States
ContactUsLinkComponent
Human Recombinant EGFR Protein (from Mammalian cells), Unconjugated
Human Recombinant EGFR Protein (from Mammalian cells), Unconjugated
Supplier:  Biortus
Certificates
undefined
Human Recombinant EGFR Protein (from Mammalian cells), Unconjugated
Supplier:  Biortus

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Mammalian cells
  • Species:
    Human
  • Gene ID:
    UniProt P00533
  • Protein/Peptide Name:
    EGFR
  • Purity:
    95%
  • Sequence:
    L25-A1210 end, C775S, C781S, C797S, C1049A, C1058A, C1146A
  • Formulation:
    50 mM Tris-HCl pH 7.5, 150 mM NaCl, 0.02% (w/v) DDM, 0.002% (w/v) CHS.
  • Shipping Temperature:
    Frozen liquid with dry ice

Specifications

Frequently Bought Together