Human Recombinant PTK6 Protein (from Insect cells), His/GST
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:His/GST
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt Q13882
- Protein/Peptide Name:PTK6
- Purity:90%
- Sequence:V2-T451
- Formulation:20 mM Tris-HCl (pH 7.5), 150 mM NaCl, 5% glycerol, 5 mM DTT, 0.1 M Trehalose
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together