Human Recombinant PGK1 Protein (from E. coli), Unconjugated
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt P00558
- Protein/Peptide Name:PGK1
- Purity:90%
- Sequence:S2-I417
- Formulation:20 mM Tris-HCl (pH 7.0)
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together