Order Entry
United States
ContactUsLinkComponent
Human Recombinant LTK Protein (from Insect cells), GST
Human Recombinant LTK Protein (from Insect cells), GST
Supplier:  Biortus
Certificates
undefined
Human Recombinant LTK Protein (from Insect cells), GST
Supplier:  Biortus

Specifications

  • Conjugation:
    GST
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Insect cells
  • Species:
    Human
  • Gene ID:
    UniProt P29376
  • Protein/Peptide Name:
    LTK
  • Purity:
    90%
  • Sequence:
    K450-S864
  • Formulation:
    50 mM Tris-HCl (pH 7.5), 500 mM NaCl, 20% glycerol, 5 mM DTT, 0.1 M Trehalose
  • Shipping Temperature:
    Frozen liquid with dry ice

Specifications

Frequently Bought Together