Human Recombinant HIPK1 Protein (from Insect cells), Unconjugated
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt Q86Z02
- Protein/Peptide Name:HIPK1
- Purity:90%
- Sequence:T158-I555
- Formulation:50 mM Tris-HCl (pH 7.5), 500 mM NaCl, 1 mM DTT, 5% glycerol
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together