Human Recombinant CSF1R Protein (from Insect cells), GST
Supplier: Biortus
Certificates
About this item
Delivery information: Frozen liquid on dry ice.
Specifications
- Conjugation:GST
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt P07333
- Protein/Peptide Name:CSF1R
- Purity:90%
- Sequence:Y538-C972
- Formulation:50 mM Tris-HCl (pH 7.5), 150 mM NaCl, 20% glycerol, 5 mM DTT, 0.1 M Trehalose
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together