Human Recombinant DCAF16/DDB1 (from Insect cells), Unconjugated
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt Q9NXF7/Q16531
- Protein Synonyms:DDB1- and CUL4-associated factor 16
- Protein/Peptide Name:DCAF16/DDB1
- Purity:90%
- Sequence:G2-L216 end/M1-H1140 end
- Formulation:20 mM HEPES (pH 7.5), 100 mM NaCl, 20% glycerol, 1 mM DTT
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together