Human Recombinant UBE2B (from Insect cells), Unconjugated
: Biortus
: Frozen liquid on dry ice.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt P63146
- Protein Synonyms:HR6B|hHR6B|E2 ubiquitin-conjugating enzyme
- Protein/Peptide Name:UBE2B
- Purity:85%
- Sequence:S2-S152
- Formulation:50 mM Tris-HCl (pH7.5), 200 mM NaCl, 20% glycerol, 1 mM DTT
- Shipping Temperature:Frozen liquid with dry ice
Frequently Bought Together