Human Recombinant UBE2V1 (from Insect cells), Unconjugated
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt Q13404
- Protein Synonyms:CROC-1|UEV-1|E2 ubiquitin-conjugating enzyme
- Protein/Peptide Name:UBE2V1
- Purity:85%
- Sequence:A2-N147
- Formulation:50 mM Tris-HCl (pH7.5), 200 mM NaCl, 5% glycerol, 1 mM DTT
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together