Human Recombinant UBE2L5 (from Insect cells), Unconjugated
Supplier: Biortus
Certificates
About this item
Delivery information: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt A0A1B0GUS4
- Protein Synonyms:E2 ubiquitin-conjugating enzyme
- Protein/Peptide Name:UBE2L5
- Purity:85%
- Sequence:A2-D154
- Formulation:50 mM Tris-HCl (pH7.5), 200 mM NaCl, 5% glycerol, 1 mM DTT
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together