Human Recombinant UBE2L3 (from Insect cells), Unconjugated
Supplier: Biortus
Certificates
About this item
Delivery information: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt P68036
- Protein Synonyms:UbcH7|E2 ubiquitin-conjugating enzyme|L-UBC
- Protein/Peptide Name:UBE2L3
- Purity:85%
- Sequence:A2-D154
- Formulation:50 mM Tris-HCl (pH7.5), 200 mM NaCl, 20% glycerol, 1 mM DTT
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together