Human Recombinant USP40 (from Mammalian cells), His/Flag
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:His/Flag
- Protein/Peptide Type:Recombinant
- Source:Mammalian cells
- Species:Human
- Gene ID:UniProt Q9NVE5
- Protein/Peptide Name:USP40
- Purity:85%
- Sequence:F2-R1235
- Formulation:50 mM Tris-HCl (pH 7.5), 200 mM NaCl, 20% glycerol, 1 mM DTT
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together