Human Recombinant USP16 (from Insect cells), Unconjugated
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt Q9Y5T5
- Protein Synonyms:Ubiquitin carboxyl-terminal hydrolase 16
- Protein/Peptide Name:USP16
- Purity:80%
- Sequence:G2-L823
- Formulation:50 mM Tris-HCl (pH 7.5), 150 mM NaCl, 5% glycerol, 2 mM DTT
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together