Human Recombinant UCHL1 (from E. coli), His
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:His
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt P09936
- Protein/Peptide Name:UCHL1
- Purity:90%
- Sequence:Q2-A223 end
- Formulation:20 mM HEPES (pH 7.5), 150 mM NaCl, 5% glycerol, 1 mM DTT
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together