Order Entry
United States
ContactUsLinkComponent
Human Recombinant STAMBP (from E. coli), Unconjugated
Human Recombinant STAMBP (from E. coli), Unconjugated
Supplier:  Biortus
Certificates
undefined
Human Recombinant STAMBP (from E. coli), Unconjugated
Supplier:  Biortus
Certificates

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Gene ID:
    UniProt O95630
  • Protein/Peptide Name:
    STAMBP
  • Purity:
    85%
  • Sequence:
    S2-R424 end
  • Formulation:
    150 mM ammonium acetate (pH 7.4)
  • Shipping Temperature:
    Frozen liquid with dry ice

Specifications

Related Information

Frequently Bought Together