Order Entry
United States
ContactUsLinkComponent
Human Recombinant CDK2/CCND1 (from Insect cells), GST/Flag/His
Human Recombinant CDK2/CCND1 (from Insect cells), GST/Flag/His
Supplier:  Biortus
Certificates
undefined
Human Recombinant CDK2/CCND1 (from Insect cells), GST/Flag/His
Supplier:  Biortus

Specifications

  • Conjugation:
    GST/Flag/His
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Insect cells
  • Species:
    Human
  • Gene ID:
    UniProt P24941/P24385
  • Protein Synonyms:
    CDK
  • Protein/Peptide Name:
    CDK2/CCND1
  • Purity:
    90%
  • Sequence:
    M1-L298 end/M1-I295 end
  • Formulation:
    50 mM Tris-HCl (pH 7.5), 150 mM NaCl, 5% glycerol
  • Shipping Temperature:
    Frozen liquid on dry ice

Specifications

Frequently Bought Together