Human Recombinant CDK19/CCNC/MED12 (from Insect cells), GST/Flag/His
: Biortus
: Frozen liquid on dry ice.
- Conjugation:GST/Flag/His
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt Q9BWU1/P24863/Q93074
- Protein Synonyms:CDK|MED12
- Protein/Peptide Name:CDK19/CCNC/MED12
- Purity:80%
- Sequence:M1-Y502/M1-S283/M1-D100
- Formulation:50 mM HEPES (pH 7.5), 150 mM NaCl, 5% glycerol
- Shipping Temperature:Frozen liquid on dry ice
Frequently Bought Together