Order Entry
United States
ContactUsLinkComponent
Human Recombinant CDK12/CCNK (from Insect cells), GST/Flag
Human Recombinant CDK12/CCNK (from Insect cells), GST/Flag
Supplier:  Biortus
Certificates
undefined
Human Recombinant CDK12/CCNK (from Insect cells), GST/Flag
Supplier:  Biortus

Specifications

  • Conjugation:
    GST/Flag
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Insect cells
  • Species:
    Human
  • Gene ID:
    UniProt Q9NYV4/O75909
  • Protein Synonyms:
    CDK
  • Protein/Peptide Name:
    CDK12/CCNK
  • Purity:
    90%
  • Sequence:
    Q696-S1082/M1-S300
  • Formulation:
    20 mM HEPES (pH 7.5), 200 mM NaCl, 5% glycerol, 1 mM DTT
  • Shipping Temperature:
    Frozen liquid on dry ice

Specifications

Related Information

Frequently Bought Together