Human Recombinant CDK1/CCNE1 (from Insect cells), GST/His
Supplier: Biortus
Certificates
About this item
Delivery information: Frozen liquid on dry ice.
Specifications
- Conjugation:GST/His
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt P06493/P24864
- Protein Synonyms:CDK
- Protein/Peptide Name:CDK1/CCNE1
- Purity:90%
- Sequence:M1-M297/M1-E410
- Formulation:20 mM HEPES (pH 7.5), 200 mM NaCl, 5% glycerol, 1 mM DTT
- Shipping Temperature:Frozen liquid on dry ice
Specifications
Frequently Bought Together