Human Recombinant CDK1/CCNB1 (from Insect cells), GST/Flag/His
: Biortus
: Frozen liquid on dry ice.
- Conjugation:GST/Flag/His
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt P06493/P14635
- Protein Synonyms:CDK
- Protein/Peptide Name:CDK1/CCNB1
- Purity:90%
- Sequence:M1-M297/N165-V433
- Formulation:50 mM Tris-HCl (pH7.5), 200 mM NaCl, 20% glycerol, 1 mM DTT
- Shipping Temperature:Frozen liquid on dry ice
Frequently Bought Together