Human Recombinant CDK1/CCNA2 (from Insect cells), Unconjugated
Supplier: Biortus
Certificates
About this item
Delivery information: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt P06493/P20248
- Protein Synonyms:CDK
- Protein/Peptide Name:CDK1/CCNA2
- Purity:85%
- Sequence:M1-M297/E174-L432
- Formulation:20 mM Tris-HCl (pH 7.5), 200 mM NaCl, 20% glycerol, 1 mM DTT
- Shipping Temperature:Frozen liquid on dry ice
Specifications
Frequently Bought Together