Human Recombinant ABCC4 (from Mammalian cells), Unconjugated
: Biortus
: Frozen liquid on dry ice.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Mammalian cells
- Species:Human
- Gene ID:UniProt O15439
- Protein Synonyms:ABC
- Protein/Peptide Name:ABCC4
- Purity:85%
- Sequence:M1-L1325 end, E1202Q
- Formulation:50 mM Tris-HCl (pH 8.5), 150 mM KCl, 2 mM DTT, 2 mM MgCl₂, 0.005% (w/v) GDN
- Shipping Temperature:Frozen liquid with dry ice
Frequently Bought Together