ContactUsLinkComponent
Eisco® Slotted Mass Set with Hanger, Stainless Steel, 9 Weights, 100 g
  470343-878
 :  Wards
undefined
Eisco® Slotted Mass Set with Hanger, Stainless Steel, 9 Weights, 100 g
  470343-878
 :  Wards
 :  PH0258OSS
  • Description:
    Slotted mass set with hanger, 100 g
  • Cat. No.:
    470343-878
Set includes nine weights totaling 100 g, made of stainless steel for durability. Set includes a hanger for suspending.
  • Stainless steel hanger with hook end for suspending masses in force experiments
  • Masses can be used for studying pressure, strength, or torque in the classroom or lab
  • Masses can be used for calibrating scales and balances
Eisco Labs stainless steel slotted masses set can be used with hanging balances as well as for calibration and studying pressure, strength, or torque in the classroom or lab.