ContactUsLinkComponent
Eisco® Hooked Stainless Steel Weights, Set of 9
Eisco® Hooked Stainless Steel Weights, Set of 9
Catalog # 470344-170
Supplier:  Wards
undefined
Eisco® Hooked Stainless Steel Weights, Set of 9
Catalog # 470344-170
Supplier:  Wards
Supplier Number:  PH0037DSS
Specifications
  • Description:
    Hooked stainless steel weights, set of 9
  • Cat. No.:
    470344-170
Specifications
About this item
Stainless steel hooked weights set with a capacity of 1000 g. Ideal for classroom or lab experiments.
  • Stainless steel weights
  • Premium wooden block
  • Weights have hooks above and below main body, but can sit flat on a surface due to retractable bottom hook
  • Will last for decades of lab and classroom use
Eisco Labs set of nine steel hooked weights, 10 to 1000 g in wooden block. Precisely machined hooked weights. Set contains (1) 1000 g, (1) 500 g, (2) 200 g, (1) 100 g, (1) 50 g, (2) 20 g, (1) 10 g.