ContactUsLinkComponent
Lumidox® II Controller
Lumidox® II Controller
Catalog # MSPP-LUM2CON
Lumidox® II Controller
Catalog # MSPP-LUM2CON
Supplier Number:  LUM2CON
Specifications
  • Description:
    Lumidox® II LED controller
  • Cat. No.:
    MSPP-LUM2CON
  • Electrical:
    100/240 V, 50/60 Hz, 3.3 A
  • Includes:
    Controller, power supply, power cable, manual
Specifications
About this item
Lumidox II Controllers are manufactured in-house. Optical power levels are provided in easy to read, easy to calculate, whole numbers. Simply scroll through the menu to select your desired output. An onboard illumination elapsed timer is provided, as well as a countdown to automatic shut off timer.
  • LED Arrays and LumLamps are custom tuned in-house
  • Provide 5 individually calibrated and configured levels of optical power
  • Arrays and LumLamps are not unique to controllers
Their calibration data is stored onboard, allowing users to use different arrays or LumLamps with one controller (only one device at a time per controller).