To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:HIAT1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:100 µl
- Epitope:1-50/490
- Form:Liquid
- Gene ID:64645
- Antigen Synonyms:MGC144858|HIAT1_HUMAN|Hippocampus abundant gene transcript 1|Tetracycline transporter like prot|Hippocampus abundant transcript|Putative tetracycline transporter like protein|Hippocampus abundant transcript 1|rCG_28876|DKFZp564L0864
- Modification:Unmodified
- Storage Buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol.
- Molecular Weight:53 kDa
- Storage Temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human HIAT1
- Tested Applications:ICC
- Purification:Purified by Protein A
- Cat. No.:76108-040
- Packaging:Vial
Specifications
About this item
The Major facilitator superfamily consists of presumed carbohydrate transporters with 10-12 membrane-spanning domains. Belonging to the facilitator superfamily, HIAT1 is a 490 amino acid multi-pass membrane protein that may function as a sugar transporter and is expressed in adult and embryonic brain. The HIAT1 gene was first observed while analyzing for active genes in neonatal mouse hippocampus. The gene encoding HIAT1 maps to human chromosome 1, the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3,000 genes on chromosome 1, and considering the great number of genes there are also a large number of diseases associated with chromosome 1. Notably, the rare aging disease Hutchinson-Gilford progeria is associated with the LMNA gene which encodes lamin A. Stickler syndrome, Parkinsons, schizophrenia, familial adenomatous polyposis, Gaucher disease and Usher syndrome are also associated with chromosome 1.
Type: Primary
Antigen: HIAT1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1-50/490
Host: Rabbit
Isotype: IgG
Reactivity: Mouse