Anti-ABCF1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # 76098-512
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:ABCF1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Western Blot:Yes
- Size:100 µl
- Epitope:501-600/845
- Form:Liquid
- Gene ID:23
- Antigen Synonyms:ATP-binding cassette sub-family F member 1|ATP-binding cassette 50|ABC50|TNF-alpha-stimulated ABC protein|ABCF1|ABC27
- Modification:Unmodified
- Storage Buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7,4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
- Molecular Weight:96 kDa
- Storage Temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze–thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human ABCF1
- Tested Applications:ICC
- Purification:Purified by Protein A
- Cat. No.:76098-512
- Packaging:Vial
Specifications
About this item
Isoform 2 is required for efficient Cap- and IRES-mediated mRNA translation initiation. Isoform 2 is not involved in the ribosome biogenesis.
Type: Primary
Antigen: ABCF1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 501-600/845
Host: Rabbit
Isotype: IgG
Reactivity: Rat