Specifications
- Antibody Type:Primary
- Antigen Name:MAPK11 Thr180+Tyr182
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:5600
- Antigen Synonyms:Mitogen activated protein kinase p38 2|MAP kinase p38 beta|MAPK 11|Mitogen activated protein kinase 11|Human p38Beta MAP kinase mRNA complete cds|MAP kinase 11|PRKM11|SAPK2|Mapk11|p38-2|protein kinase mitogen activated 11|P38B|Stress activated protein kinase 2|P38BETA2|mitogen-activated protein kinase 11|p38 2|SAPK2B|Mitogen activated protein kinase p38 beta|MK11_HUMAN|MAPK11phospho T180/Y182|Mitogen-activated protein kinase p38 beta|p38Beta|Stress-activated protein kinase 2
- Modification:Phosphorylation (Thr180+Tyr182)
- Storage Buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
- Molecular Weight:41 kDa
- Storage Temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic phosphopeptide derived from human MAPK11 around the phosphorylation site of Thr180+Tyr182
- Tested Applications:ICC
- Purification:Purified by Protein A
- Cat. No.:76080-616
- Packaging:Vial
Specifications
About this item
The protein encoded by this gene is a member of the MAP kinase family. MAP kinases act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation, and development. MAPK11 is most closely related to p38 MAP kinase, both of which can be activated by proinflammatory cytokines and environmental stress. This kinase is activated through its phosphorylation by MAP kinase kinases (MKKs), preferably by MKK6. Transcription factor ATF2/CREB2 has been shown to be a substrate of this kinase.
Type: Primary
Antigen: MAPK11 Thr180+Tyr182
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human