Anti-ABAT Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # 76078-236
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:ABAT
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µl
- Epitope:151-250/500
- Form:Liquid
- Gene ID:18
- Antigen Synonyms:GABA-AT|(S)-3-amino-2-methylpropionate transaminase|NPD009|L-AIBAT|GABA-T|GABA aminotransferase|4-aminobutyrate aminotransferase, mitochondrial|ABAT|Gamma-amino-N-butyrate transaminase|GABA transaminase|GABAT
- Modification:Unmodified
- Storage Buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
- Molecular Weight:56 kDa
- Storage Temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human ABAT
- Tested Applications:ICC
- Purification:Purified by Protein A
- Cat. No.:76078-236
- Packaging:Vial
Specifications
About this item
Catalyzes the conversion of gamma-aminobutyrate and L-beta-aminoisobutyrate to succinate semialdehyde and methylmalonate semialdehyde, respectively. Can also convert delta-aminovalerate and beta-alanine.
Type: Primary
Antigen: ABAT
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 151-250/500
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat