Anti-FANCG Rabbit Polyclonal Antibody (Alexa Fluor® 750)
76077-576
: Bioss
- Antibody Type:Primary
- Antigen Name:FANCG
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Epitope:13-110/622
- Form:Liquid
- Gene ID:2189
- Antigen Synonyms:Protein FACG|DNA repair protein XRCC9|FAG|FANCG_HUMAN|X ray repair, complementing defective, in Chinese hamster cells 9|XRCC9|Fanconi anaemia complementation group G|DNA-repair protein XRCC9
- Modification:Unmodified
- Storage Buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
- Molecular Weight:69 kDa
- Storage Temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human FANCG
- Tested Applications:ICC
- Purification:Purified by Protein A
- Cat. No.:76077-576
- Packaging:Vial
FANCG, involved in Fanconi anemia, confers resistance to both hygromycin and mitomycin C. FANCG contains a 5-prime GC-rich untranslated region characteristic of housekeeping genes. The putative 622-amino acid protein has a leucine-zipper motif at its N-terminus. Fanconi anemia is an autosomal recessive disorder with diverse clinical symptoms, including developmental anomalies, bone marrow failure, and early occurrence of malignancies. A minimum of 8 FA genes have been identified.
Type: Primary
Antigen: FANCG
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 13-110/622
Host: Rabbit
Isotype: IgG
Reactivity: Human