Anti-Quiescin Q6 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # 76118-214
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:Quiescin Q6
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Size:100 µl
- Epitope:244-350/747
- Form:Liquid
- Gene ID:5768
- Antigen Synonyms:QSOX1|Quiescin Q6 sulfhydryl oxidase 1|Skin sulfhydryl oxidase|Sulfhydryl oxidase 1|Human quiescin Q6 mRNA, partial cds|Sox|Sulfhydryl oxidase 1 precursor|Q6 antibody QSCN6|QSOX1_HUMAN|Quiescin Q6|hQSOX
- Modification:Unmodified
- Storage Buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol.
- Molecular Weight:80 kDa
- Storage Temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Quiescin Q6/QSOX1
- Tested Applications:ICC
- Purification:Purified by Protein A
- Cat. No.:76118-214
- Packaging:Vial
Specifications
About this item
This gene encodes a protein that contains domains of thioredoxin and ERV1, members of two long-standing gene families. The gene expression is induced as fibroblasts begin to exit the proliferative cycle and enter quiescence, suggesting that this gene plays an important role in growth regulation. Two transcript variants encoding two different isoforms have been found for this gene.
Type: Primary
Antigen: Quiescin Q6
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 244-350/747
Host: Rabbit
Isotype: IgG
Reactivity: