Anti-LETMD1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # 76081-392
Supplier: Bioss
Specifications
- Antibody Type:Primary
- Antigen Name:LETMD1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western Blot:Yes
- Size:100 µl
- Epitope:21-120/360
- Form:Liquid
- Gene ID:25875
- Antigen Synonyms:HCCR 2|HCCR-1|HCCR2|Cervical cancer proto-oncogene 2 protein|LETM1 domain-containing protein 1|Cervical cancer 1 proto-oncogene protein p40|LTMD1_HUMAN|HCRR-2
- Modification:Unmodified
- Storage Buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol.
- Molecular Weight:40 kDa
- Storage Temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human LETMD1
- Tested Applications:ICC
- Purification:Purified by Protein A
- Cat. No.:76081-392
- Packaging:Vial
Specifications
About this item
This gene encodes a mitochondrial outer membrane protein. It has a potential role in tumorigenesis, which may result from negative regulation of the p53 tumor suppressor gene. Alternatively spliced transcript variants have been noted for this gene.
Type: Primary
Antigen: LETMD1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 21-120/360
Host: Rabbit
Isotype: IgG
Reactivity: