Anti-GPCR HM74 Rabbit Polyclonal Antibody (Cy5)
10315-156
: Bioss
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:GPCR HM74
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:
- Size:100 µl
- Environmentally Preferable:
- Epitope:51-150/387
- Form:Liquid
- Antigen Synonyms:NIACR2|HM74|G-protein coupled receptor HM74|GPR109B|HCAR3|G-protein coupled receptor 109B|HCA3|Nicotinic acid receptor 2|Niacin receptor 2|HM74B|G-protein coupled receptor HM74B|Hydroxycarboxylic acid receptor 3|Puma-g|HCAR3_HUMAN|PUMAG
- Modification:Unmodified
- Storage Buffer:Aqueous buffered solution containing 0.01 M TBS (pH 7.4) with 1% BSA, 0.03% Proclin300 and 50% Glycerol
- Molecular Weight:44 kDa
- Storage Temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping Temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human GPCR HM74
- Tested Applications:ICC
- Purification:Purified by Protein A
- Cat. No.:10315-156
- Packaging:Vial
GPCR HM74.
Type: Primary
Antigen: GPCR HM74
Clonality: Polyclonal
Clone:
Conjugation: Cy5
Public Immunogen Range: 51-150/387
Host: Rabbit
Isotype: IgG
Reactivity: