Lighthouse Chemo Bio-Wipe Bag
Catalog # 76716-820
Supplier: Unimed-Midwest
Specifications
- Color:Yellow
- Description:Chemo bio-wipe bag
- Cat. No.:76716-820
- W×L:11.75×12.13"
Specifications
About this item
Quick, economical cleanup for biohazardous Chemo spills.
- Provides a convenient, temporary containment vehicle for proper disposal into an approved OSHA/ASHP container
- Reduces risk of contact with harmful chemotherapy drugs, when used with chemo-tested medical exam gloves
- Complies with OSHA Standard 29CFR 1910.1030 for PPE
- Composed of a patented chemo yellow opaque polyethylene plastic bag on one side, and an absorbable pad bonded to the other side
The absorbent pad (11.75×9") capacity is approximately 1/2 pint. An adhesive strip provides closure for safe containment and proper disposal, and an ambidextrous mitt is included inside the bag to provide better maneuverability. Made in the USA.