L(+)-Histidine monohydrochloride monohydrate, Multi-Compendial, J.T.Baker®
Supplier: Avantor Performance Materials
Certificates
About this item
CAS Number:5934-29-2
:
Formula:C₆H₉N₃O₂·HCl·H₂O
MDL Number:MFCD00151027
:209.63 g/mol
Merck Index:13,04741
Melting Pt:259 °C
Density:1.485 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:211-438-9
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Assay (dried substance) [Ph.Eur./BP] :98.5 - 101.0 %
- Identification A :Passes test Ph.Eur./BP
- Identification B :Passes test Ph.Eur./BP
- Identification C :Passes test Ph.Eur./BP
- Identification F :Passes test Ph.Eur./BP
- Appearance of solution:Passes test Ph.Eur./BP
- Spec. opt. rot. (on dried) [Ph.Eur./BP]:9.2 - 10.6 °
- Loss on drying (145-150°C) [Ph.Eur./BP] :7.0 - 10.0 %
- pH [Ph.Eur./BP] :3.0 - 5.0
- SO₄ (Sulphate) [Ph.Eur./BP] :≤ 300 ppm
- Fe (Iron) [Ph.Eur./BP] :≤ 10 ppm
- Ninhydrin-pos.subst.(each) [Ph.Eur./BP] :≤ 0.2 %
- Ninhydrin-pos.subst.(total) [Ph.Eur./BP] :≤ 0.5 %
- Ammonium (570 nm) [Ph.Eur./BP] :≤ 0.02 %
- Sulphated ash [Ph.Eur./BP] :≤ 0.1 %
- Assay (calculated on anhydrous) [JP] :99.0 - 101.0 %
- Identification:Passes test JP
- Specific optical rotation [JP] :9.2 - 10.6 °
- pH [JP] :3.5 - 4.5
- Clarity and color of solution [JP] :Passes test JP
- SO₄ (Sulphate) [JP] :≤ 0.028 %
- NH₄ (Ammonium) [JP] :≤ 0.02 %
- Fe (Iron) [JP] :≤ 10 ppm
- Related substances :Passes test JP
- Water [JP] :7.2 - 10.0 %
- Residue on ignition [JP] :≤ 0.1 %
- Total aerobic microbial count :≤ 10 CFU/g
- Total mold and yeast count :≤ 10 CFU/g
- Endotoxin :≤ 10 EU/g
Specification Test Results
Related Information
Frequently Bought Together